Methyl 3-bromo-2,4-difluorobenzoate 95%
Leverantör: Apollo Scientific
Varning
PC210039-500MG PC210039-1G PC210039-250MG
APOSPC210039-500MGEA 9075 SEK
APOSPC210039-500MG APOSPC210039-1G APOSPC210039-250MG
Methyl 3-bromo-2,4-difluorobenzoate 95%
Methyl 3-bromo-2,4-difluorobenzoate
Formel: C₈H₅BrF₂O₂
MW: 251,025 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09261258
CAS nummer: 935534-49-9