5-Fluoro-1H-indole-3-carboxylic acid 97%
Leverantör: Apollo Scientific
PC201206-10G PC201206-1G PC201206-5G
APOSPC201206-10GEA 8275 SEK
APOSPC201206-10G APOSPC201206-1G APOSPC201206-5G
5-Fluoro-1H-indole-3-carboxylic acid 97%
5-Fluoro-1H-indole-3-carboxylic acid
Formel: C₉H₆FNO₂
MW: 179,148 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00047171
CAS nummer: 23077-43-2