4-Bromo-6-fluoro-2,3-dihydro-1H-inden-1-one 95+%

Leverantör: Apollo Scientific

PC201173-100MG PC201173-1G PC201173-250MG
APOSPC201173-100MGEA 995 SEK
APOSPC201173-100MG APOSPC201173-1G APOSPC201173-250MG
4-Bromo-6-fluoro-2,3-dihydro-1H-inden-1-one 95+%
4-Bromo-6-fluoro-2,3-dihydro-1H-inden-1-one
Formel: C₉H₆BrFO
MW: 229,046 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09038174
CAS nummer: 174603-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR