2-Chloro-5-fluoro-4-methylpyrimidine 98%
Leverantör: Apollo Scientific
PC201121-100MG PC201121-1G PC201121-250MG
APOSPC201121-100MGEA 1150 SEK
APOSPC201121-100MG APOSPC201121-1G APOSPC201121-250MG
2-Chloro-5-fluoro-4-methylpyrimidine 98%
2-Chloro-5-fluoro-4-methylpyrimidine
Formel: C₅H₄ClFN₂
MW: 146,55 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD14584604
CAS nummer: 134000-96-7