(R)-2-(2,5-Difluorophenyl)pyrrolidine hydrochloride 95+%
Leverantör: Apollo Scientific
PC201077-100MG PC201077-1G PC201077-250MG
APOSPC201077-100MGEA 600 SEK
APOSPC201077-100MG APOSPC201077-1G APOSPC201077-250MG
(R)-2-(2,5-Difluorophenyl)pyrrolidine hydrochloride 95+%
(R)-2-(2,5-Difluorophenyl)pyrrolidine hydrochloride
Formel: C₁₀H₁₁F₂N·ClH
MW: 219,659 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08751413
CAS nummer: 1218935-60-4