Ethyl 3-fluoro-2-methylbenzoate 98%

Leverantör: Apollo Scientific

PC201026-10G PC201026-1G PC201026-5G
APOSPC201026-10GEA 4725 SEK
APOSPC201026-10G APOSPC201026-1G APOSPC201026-5G
Ethyl 3-fluoro-2-methylbenzoate 98%
Ethyl 3-fluoro-2-methylbenzoate
Formel: C₁₀H₁₁FO₂
MW: 182,192 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06204676
CAS nummer: 114312-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR