3-Bromo-2-(2-fluorophenoxy)pyridine 97%
Leverantör: Apollo Scientific
Varning
PC200389-1G PC200389-500MG
APOSPC200389-1GEA 7900 SEK
APOSPC200389-1G APOSPC200389-500MG
3-Bromo-2-(2-fluorophenoxy)pyridine 97%
3-Bromo-2-(2-fluorophenoxy)pyridine
Formel: C₁₁H₇BrFNO
MW: 268.082 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD14651747
CAS nummer: 1215809-12-3