Triphenyl(2,2,2-trifluoroethyl)phosphanium trifluoromethanesulphonate 97%
Leverantör: Apollo Scientific
Varning
PC200031-1G PC200031-5G
APOSPC200031-1GEA 3025 SEK
APOSPC200031-1G APOSPC200031-5G
Triphenyl(2,2,2-trifluoroethyl)phosphanium trifluoromethanesulphonate 97%
Triphenyl(2,2,2-trifluoroethyl)phosphanium trifluoromethanesulphonate
Formel: C₂₀H₁₇F₃P·CF₃O₃S
MW: 494,387 g/mol
Smältpunkt: 208…209 °C
Storage Temperature: Omgivande
MDL Number: MFCD22689151
CAS nummer: 135654-55-6