Pentafluorophenyl 4-morpholin-4-ylbenzoate 97%
Leverantör: Apollo Scientific
PC1228-1G PC1228-250MG
APOSPC1228-1GEA 2600 SEK
APOSPC1228-1G APOSPC1228-250MG
Pentafluorophenyl 4-morpholin-4-ylbenzoate 97%
Pentafluorophenyl 4-morpholin-4-ylbenzoate
Formel: C₁₇H₁₂F₅NO₃
MW: 373.274 g/mol
Smältpunkt: 144…146.5 °C
Storage Temperature: Omgivande
MDL Number: MFCD09817462
CAS nummer: 921938-51-4