3-(3-Fluorophenyl)prop-2-yn-1-ol 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-(3-Fluorophenyl)-3-hydroxyprop-1-yne
PC11155-1G PC11155-250MG PC11155-5G
APOSPC11155-1GEA 2925 SEK
APOSPC11155-1G APOSPC11155-250MG APOSPC11155-5G
3-(3-Fluorophenyl)prop-2-yn-1-ol 97%
3-(3-Fluorophenyl)prop-2-yn-1-ol
Formel: C₉H₇FO
MW: 150,15 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD04039231
CAS nummer: 197239-54-6