3-(2-Fluorophenyl)prop-2-yn-1-ol 97%
Leverantör: Apollo Scientific
Varning
PC11154-1G PC11154-250MG PC11154-5G
APOSPC11154-1GEA 1400 SEK
APOSPC11154-1G APOSPC11154-250MG APOSPC11154-5G
3-(2-Fluorophenyl)prop-2-yn-1-ol 97%
3-(2-Fluorophenyl)prop-2-yn-1-ol
Formel: C₉H₇FO
MW: 150,15 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD04039230
CAS nummer: 208183-93-1