Ethyl 8-bromo-5-fluoro-4-hydroxyquinoline-3-carboxylate
Leverantör: Apollo Scientific
PC110038-1G PC110038-250MG
APOSPC110038-1GEA 3350 SEK
APOSPC110038-1G APOSPC110038-250MG
Ethyl 8-bromo-5-fluoro-4-hydroxyquinoline-3-carboxylate
Ethyl 8-bromo-5-fluoro-4-hydroxyquinoline-3-carboxylate
Formel: C₁₂H₉BrFNO₃
MW: 314.107 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16036881
CAS nummer: 655236-28-5