2-(Aminomethyl)-3-fluoro-6-methylpyridine hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: (3-Fluoro-6-methylpyridin-2-yl)methylamine hydrochloride, (3-Fluoro-6-methylpyridin-2-yl)methanamine hydrochloride
PC10659-1G PC10659-250MG
APOSPC10659-1GEA 9000 SEK
APOSPC10659-1G APOSPC10659-250MG
2-(Aminomethyl)-3-fluoro-6-methylpyridine hydrochloride
2-(Aminomethyl)-3-fluoro-6-methylpyridine hydrochloride
Formel: C₇H₉FN₂·ClH
MW: 176.619 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD17214195
CAS nummer: 1257535-11-7