(2R)-2-(Trifluoromethyl)pyrrolidine 95%
Leverantör: Apollo Scientific
|
Fara
|
Synonyms:
2(R)-2-Trifluoromethylpyrrolidine
PC1059-1G
PC1059-5G
APOSPC1059-1GEA
3025
SEK
APOSPC1059-1G
APOSPC1059-5G
(2R)-2-(Trifluoromethyl)pyrrolidine 95%
(2R)-2-(Trifluoromethyl)pyrrolidine
Formel:
C₅H₈F₃N MW: 139.119 g/mol Kokpunkt: 110…112 °C Smältpunkt: 29…33 °C Flampunkt: 28.9 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD07784180 CAS nummer: 1073556-31-6 UN: 2925 ADR: 4.1+8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...