Perfluoro(4-methyl-3,6-dioxaoctane)sulphonyl fluoride 97%
Leverantör: Apollo Scientific
PC1023-1G PC1023-5G
APOSPC1023-1GEA 1375 SEK
APOSPC1023-1G APOSPC1023-5G
Perfluoro(4-methyl-3,6-dioxaoctane)sulphonyl fluoride 97%
Perfluoro(4-methyl-3,6-dioxaoctane)sulphonyl fluoride
Formel: C₇F₁₆O₄S
MW: 484.112 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00155782
CAS nummer: 27744-59-8