2-[(2-Fluorophenyl)amino]benzoic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-(2-Fluoroanilino)benzoic acid, N-(2-Fluorophenyl)anthranilic acid
PC0937-1G PC0937-5G
APOSPC0937-1GEA 210 SEK
APOSPC0937-1G APOSPC0937-5G
2-[(2-Fluorophenyl)amino]benzoic acid 98%
2-[(2-Fluorophenyl)amino]benzoic acid
Formel: C₁₃H₁₀FNO₂
MW: 231,222 g/mol
Smältpunkt: 186 °C
Storage Temperature: Omgivande
MDL Number: MFCD00662257
CAS nummer: 54-58-0