(3R)-3-Hydroxy-4,4,4-trifluorobutanoic acid 97%
Leverantör: Apollo Scientific
Fara
Synonyms: (3R)-3-Hydroxy-4,4,4-trifluorobutyric acid
PC0620-250MG PC0620-100MG PC0620-1G
APOSPC0620-250MGEA 3875 SEK
APOSPC0620-250MG APOSPC0620-100MG APOSPC0620-1G
(3R)-3-Hydroxy-4,4,4-trifluorobutanoic acid 97%
(3R)-3-Hydroxy-4,4,4-trifluorobutanoic acid
Formel: C₄H₅F₃O₃
MW: 158,076 g/mol
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD03093017
CAS nummer: 108211-36-5
UN: 3261
ADR: 8,III