4,5-Difluoro-2-(trifluoromethyl)benzamide
Leverantör: Apollo Scientific
Synonyms: 2-(Aminocarbonyl)-4,5-difluorobenzotrifluoride
PC0298-1G PC0298-5G
APOSPC0298-1GEA 3525 SEK
APOSPC0298-1G APOSPC0298-5G
4,5-Difluoro-2-(trifluoromethyl)benzamide
4,5-Difluoro-2-(trifluoromethyl)benzamide
Formel: C₈H₄F₅NO
MW: 225,115 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01631486
CAS nummer: 261944-97-2