2,2-Bis(trifluoromethyl)propanoyl fluoride
Leverantör: Apollo Scientific
Fara
Synonyms: 2-Methyl-3,3,3-trifluoro-2-(trifluoromethyl)propanoyl fluoride, 2,2-Bis(trifluoromethyl)propionyl fluoride
PC0203-1G PC0203-25G PC0203-5G
APOSPC0203-1GEA 1300 SEK
APOSPC0203-1G APOSPC0203-25G APOSPC0203-5G
2,2-Bis(trifluoromethyl)propanoyl fluoride
2,2-Bis(trifluoromethyl)propanoyl fluoride
Formel: C₅H₃F₇O
MW: 212,066 g/mol
Kokpunkt: 47…49 °C
Densitet: 1,473 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00236084
CAS nummer: 1735-87-1
UN: 3265
ADR: 8,III