(1S)-(-)-1-[4-(Trifluoromethyl)phenyl]propan-1-ol 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 4-[(1S)-(-)-1-Hydroxyprop-1-yl]benzotrifluoride, (S)-(-)-α-Ethyl-4-(trifluoromethyl)benzyl alcohol
PC01651-1G PC01651-250MG
APOSPC01651-1GEA 14200 SEK
APOSPC01651-1G APOSPC01651-250MG
(1S)-(-)-1-[4-(Trifluoromethyl)phenyl]propan-1-ol 97%
(1S)-(-)-1-[4-(Trifluoromethyl)phenyl]propan-1-ol
Formel: C₁₀H₁₁F₃O
MW: 204.189 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16170458
CAS nummer: 73854-05-4