2-Bromo-6-fluorothioanisole
Leverantör: Apollo Scientific
Synonyms: 2-Bromo-6-fluorophenyl methyl sulphide, 1-Bromo-3-fluoro-2-(methylsulphanyl)benzene
PC01512-1G PC01512-25G PC01512-5G
APOSPC01512-1GEA 3250 SEK
APOSPC01512-1G APOSPC01512-25G APOSPC01512-5G
2-Bromo-6-fluorothioanisole
2-Bromo-6-fluorothioanisole
Formel: C₇H₆BrFS
MW: 221,09 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22056775
CAS nummer: 1370025-53-8