4-[4-(Ethoxycarbonyl)-1,3-thiazol-2-yl]piperidine, N-BOC protected 95%
Leverantör: Apollo Scientific
Varning
Synonyms: tert-Butyl 4-[4-(ethoxycarbonyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate
OR9717-1G OR9717-250MG
APOSOR9717-1GEA 2250 SEK
APOSOR9717-1G APOSOR9717-250MG
4-[4-(Ethoxycarbonyl)-1,3-thiazol-2-yl]piperidine, N-BOC protected 95%
4-[4-(Ethoxycarbonyl)-1,3-thiazol-2-yl]piperidine, N-BOC protected
Formel: C₁₆H₂₄N₂O₄S
MW: 340,438 g/mol
Smältpunkt: 71…73 °C
Storage Temperature: Omgivande
MDL Number: MFCD06659069
CAS nummer: 365413-31-6