4-Iodophenylhydrazine hydrochloride 97%
Leverantör: Apollo Scientific
Fara
Synonyms: (4-Iodophenyl)hydrazine hydrochloride
OR9707-25G OR9707-5G OR9707-100G
APOSOR9707-25GEA 1900 SEK
APOSOR9707-25G APOSOR9707-5G APOSOR9707-100G
4-Iodophenylhydrazine hydrochloride 97%
4-Iodophenylhydrazine hydrochloride
Formel: C₆H₇IN₂·ClH
Storage Temperature: Omgivande
MDL Number: MFCD00060724
CAS nummer: 62830-55-1
UN: 2811
ADR: 6.1,III