(7-Methoxyimidazo[1,2-a]pyridin-2-yl)-methanol 97%
Leverantör: Apollo Scientific
Varning
OR965960-1G OR965960-500MG
APOSOR965960-1GEA 11175 SEK
APOSOR965960-1G APOSOR965960-500MG
(7-Methoxyimidazo[1,2-a]pyridin-2-yl)-methanol 97%
(7-Methoxyimidazo[1,2-a]pyridin-2-yl)-methanol
Formel: C₉H₁₀N₂O₂
MW: 178.188 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD29067469
CAS nummer: 1781647-84-4