3-(2-Thiazolyl)propionic acid 95%
Leverantör: Apollo Scientific
OR964201-1G OR964201-250MG
APOSOR964201-1GEA 5150 SEK
APOSOR964201-1G APOSOR964201-250MG
3-(2-Thiazolyl)propionic acid 95%
3-(2-Thiazolyl)propionic acid
Formel: C₆H₇NO₂S
MW: 157,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02683062
CAS nummer: 144163-65-5