2-Chloroimidazo[1,2-a]pyridine 97%
Leverantör: Apollo Scientific
OR963889-1G OR963889-250MG
APOSOR963889-1GEA 7875 SEK
APOSOR963889-1G APOSOR963889-250MG
2-Chloroimidazo[1,2-a]pyridine 97%
2-Chloroimidazo[1,2-a]pyridine
Formel: C₇H₅ClN₂
MW: 152,581 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06335049
CAS nummer: 3999-05-1