2,3,4,5-Tetrahydro-benzo[c]azepin-1-one 97%
Leverantör: Apollo Scientific
Synonyms: 2,3,4,5-Tetrahydro-1H-benzo[c]azepin-1-one
OR963516-10G OR963516-1G OR963516-5G
APOSOR963516-10GEA 10950 SEK
APOSOR963516-10G APOSOR963516-1G APOSOR963516-5G
2,3,4,5-Tetrahydro-benzo[c]azepin-1-one 97%
2,3,4,5-Tetrahydro-benzo[c]azepin-1-one
Formel: C₁₀H₁₁NO
MW: 161,2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04114867
CAS nummer: 6729-50-6