trans-3-Chlorocyclobutane-1-carboxylic acid

Leverantör: Apollo Scientific

OR962962-10G OR962962-1G OR962962-5G
APOSOR962962-10GEA 20900 SEK
APOSOR962962-10G APOSOR962962-1G APOSOR962962-5G
trans-3-Chlorocyclobutane-1-carboxylic acid
trans-3-Chlorocyclobutane-1-carboxylic acid
Formel: C₅H₇ClO₂
MW: 134,561 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16661183
CAS nummer: 35207-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR