2-Methoxy-4-(pyrrolidin-1-yl)aniline 96%

Leverantör: Apollo Scientific

OR962869-1G OR962869-5G
APOSOR962869-1GEA 1975 SEK
APOSOR962869-1G APOSOR962869-5G
2-Methoxy-4-(pyrrolidin-1-yl)aniline 96%
2-Methoxy-4-(pyrrolidin-1-yl)aniline
Formel: C₁₁H₁₆N₂O
MW: 192,258 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11848953
CAS nummer: 143525-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR