N-Methoxy-6,n-dimethyl-nicotinamide 95%

Leverantör: Apollo Scientific

OR962777-1G OR962777-5G
APOSOR962777-1GEA 2750 SEK
APOSOR962777-1G APOSOR962777-5G
N-Methoxy-6,n-dimethyl-nicotinamide 95%
N-Methoxy-6,n-dimethyl-nicotinamide
Formel: C₉H₁₂N₂O₂
MW: 180,204 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09907654
CAS nummer: 221615-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR