3-(2-Methoxyethoxy)benzoic acid 95%
Leverantör: Apollo Scientific
OR962735-1G OR962735-250MG
APOSOR962735-1GEA 3350 SEK
APOSOR962735-1G APOSOR962735-250MG
3-(2-Methoxyethoxy)benzoic acid 95%
3-(2-Methoxyethoxy)benzoic acid
Formel: C₁₀H₁₂O₄
MW: 196.2 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD09036534
CAS nummer: 152808-60-1