3-Piperidinobenzaldehyde 96%

Leverantör: Apollo Scientific

OR962683-10G OR962683-1G OR962683-5G
APOSOR962683-10GEA 8875 SEK
APOSOR962683-10G APOSOR962683-1G APOSOR962683-5G
3-Piperidinobenzaldehyde 96%
3-Piperidinobenzaldehyde
Formel: C₁₂H₁₅NO
Storage Temperature: Omgivande
MDL Number: MFCD07772815
CAS nummer: 669050-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR