1-O-Tolyl-1H-pyrazole 98%
Leverantör: Apollo Scientific
Synonyms: 1-(o-Tolyl)-1H-pyrazole
OR962093-10G OR962093-1G OR962093-5G
APOSOR962093-10GEA 7625 SEK
APOSOR962093-10G APOSOR962093-1G APOSOR962093-5G
1-O-Tolyl-1H-pyrazole 98%
1-O-Tolyl-1H-pyrazole
Formel: C₁₀H₁₀N₂
MW: 158,2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12192640
CAS nummer: 20157-44-2