3-(5-Bromothien-2-yl)-D-alanine 97%
Leverantör: Apollo Scientific
OR962047-1G OR962047-500MG
APOSOR962047-1GEA 5175 SEK
APOSOR962047-1G APOSOR962047-500MG
3-(5-Bromothien-2-yl)-D-alanine 97%
3-(5-Bromothien-2-yl)-D-alanine
Formel: C₇H₈BrNO₂S
MW: 250,113 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04117831
CAS nummer: 264903-54-0