1-Methyl-4-phenyl-piperidin-4-ol 95%

Leverantör: Apollo Scientific

OR961986-1G OR961986-5G
APOSOR961986-1GEA 3600 SEK
APOSOR961986-1G APOSOR961986-5G
1-Methyl-4-phenyl-piperidin-4-ol 95%
1-Methyl-4-phenyl-piperidin-4-ol
Formel: C₁₂H₁₇NO
MW: 191,269 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00464382
CAS nummer: 4972-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR