7-Methyl-2,4-dihydro-1,4-benzoxazin-3-one 98%
Leverantör: Apollo Scientific
Synonyms: 7-Methyl-2H-benzo[b][1,4]oxazin-3(4H)-one
OR960212-1G OR960212-5G
APOSOR960212-1GEA 3175 SEK
APOSOR960212-1G APOSOR960212-5G
7-Methyl-2,4-dihydro-1,4-benzoxazin-3-one 98%
7-Methyl-2,4-dihydro-1,4-benzoxazin-3-one
Formel: C₉H₉NO₂
MW: 163,173 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03019817
CAS nummer: 39522-25-3