3-(Methylamino)benzoic acid hydrochloride 95%
Leverantör: Apollo Scientific
OR960209-1G OR960209-250MG
APOSOR960209-1GEA 1575 SEK
APOSOR960209-1G APOSOR960209-250MG
3-(Methylamino)benzoic acid hydrochloride 95%
3-(Methylamino)benzoic acid hydrochloride
Formel: C₈H₉NO₂·ClH
MW: 187.624 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09752165
CAS nummer: 1194804-60-8