4-Phenylcyclohex-3-en-1-one 98%
Leverantör: Apollo Scientific
OR960163-1G OR960163-5G
APOSOR960163-1GEA 3675 SEK
APOSOR960163-1G APOSOR960163-5G
4-Phenylcyclohex-3-en-1-one 98%
4-Phenylcyclohex-3-en-1-one
Formel: C₁₂H₁₂O
Storage Temperature: Omgivande
MDL Number: MFCD17015556
CAS nummer: 51171-71-2