2,4-Dibromo-3a,4,7,7a-tetrahydro-1H-4,7-methanoindene-1,8-dione
Leverantör: Apollo Scientific
Synonyms: 4,7-Dibromotricyclo[5.2.1.0^{2,6}]deca-4,8-diene-3,10-dione
OR960141-1G OR960141-5G
APOSOR960141-1GEA 3075 SEK
APOSOR960141-1G APOSOR960141-5G
2,4-Dibromo-3a,4,7,7a-tetrahydro-1H-4,7-methanoindene-1,8-dione
2,4-Dibromo-3a,4,7,7a-tetrahydro-1H-4,7-methanoindene-1,8-dione
Formel: C₁₀H₆Br₂O₂
MW: 317,961 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05664459
CAS nummer: 32846-64-3