2-(Isobutylamino)benzonitrile 97%
Leverantör: Apollo Scientific
OR960002-1G
OR960002-5G
APOSOR960002-1GEA
1975
SEK
APOSOR960002-1G
APOSOR960002-5G
2-(Isobutylamino)benzonitrile 97%
2-(Isobutylamino)benzonitrile
Formel:
C₁₁H₁₄N₂ MW: 174.242 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD11144635 CAS nummer: 204078-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...