5,6-Dimethyl-3H-thieno[2,3-d]pyrimidin-4-one 96%
Leverantör: Apollo Scientific
Synonyms: 5,6-Dimethylthieno[2,3-d]pyrimidin-4(3H)-one
OR959568-10G OR959568-1G OR959568-5G
APOSOR959568-10GEA 11225 SEK
APOSOR959568-10G APOSOR959568-1G APOSOR959568-5G
5,6-Dimethyl-3H-thieno[2,3-d]pyrimidin-4-one 96%
5,6-Dimethyl-3H-thieno[2,3-d]pyrimidin-4-one
Formel: C₈H₈N₂OS
MW: 180,227 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00463622
CAS nummer: 18593-44-7