(3R-Cis)-3,6-dimethyl-1,4-dioxane-2,5-dione 98%
Leverantör: Apollo Scientific
Synonyms: (3R,6R)-3,6-Dimethyl-1,4-dioxane-2,5-dione
OR959522-10G OR959522-25G OR959522-5G
APOSOR959522-10GEA 1050 SEK
APOSOR959522-10G APOSOR959522-25G APOSOR959522-5G
(3R-Cis)-3,6-dimethyl-1,4-dioxane-2,5-dione 98%
(3R-Cis)-3,6-dimethyl-1,4-dioxane-2,5-dione
Formel: C₆H₈O₄
MW: 144,125 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00082566
CAS nummer: 13076-17-0