2,3-Dihydrobenzo[b]furan-7-ylamine 95%

Leverantör: Apollo Scientific

Synonyms: 2,3-Dihydrobenzofuran-7-amine

OR958944-1G OR958944-250MG
APOSOR958944-1GEA 2100 SEK
APOSOR958944-1G APOSOR958944-250MG
2,3-Dihydrobenzo[b]furan-7-ylamine 95%
2,3-Dihydrobenzo[b]furan-7-ylamine
Formel: C₈H₉NO
MW: 135,163 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11109313
CAS nummer: 13414-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR