5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidin-2-ol hydrochloride 95%
Leverantör: Apollo Scientific
OR958464-1G OR958464-250MG
APOSOR958464-1GEA 9500 SEK
APOSOR958464-1G APOSOR958464-250MG
5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidin-2-ol hydrochloride 95%
5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidin-2-ol hydrochloride
Formel: C₇H₉N₃O·ClH
MW: 187,627 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16615181
CAS nummer: 1956321-99-5