3-Hydroxy-3-methylhexanoic acid 95%
Leverantör: Apollo Scientific
Varning
OR958055-1G OR958055-250MG
APOSOR958055-1GEA 3400 SEK
APOSOR958055-1G APOSOR958055-250MG
3-Hydroxy-3-methylhexanoic acid 95%
3-Hydroxy-3-methylhexanoic acid
Formel: C₇H₁₄O₃
MW: 146.184 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07779366
CAS nummer: 58888-76-9