3-(Methanesulphonylmethyl)benzonitrile 98%
Leverantör: Apollo Scientific
OR956195-1G
OR956195-5G
APOSOR956195-1GEA
3450
SEK
APOSOR956195-1G
APOSOR956195-5G
3-(Methanesulphonylmethyl)benzonitrile 98%
3-(Methanesulphonylmethyl)benzonitrile
Formel:
C₉H₉NO₂S MW: 195,238 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD19257809 CAS nummer: 261924-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...