5-Bromopyridine-3-sulphonic acid 96%
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Bromo-5-sulphopyridine
OR9549-1G OR9549-5G
APOSOR9549-1GEA 2325 SEK
APOSOR9549-1G APOSOR9549-5G
5-Bromopyridine-3-sulphonic acid 96%
5-Bromopyridine-3-sulphonic acid
Formel: C₅H₄BrNO₃S
MW: 238,059 g/mol
Smältpunkt: >300 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00717696
CAS nummer: 62009-34-1