2-(Cyclopropylamino)ethanol 98%
Leverantör: Apollo Scientific
Varning
OR953960-1G OR953960-250MG
APOSOR953960-1GEA 5575 SEK
APOSOR953960-1G APOSOR953960-250MG
2-(Cyclopropylamino)ethanol 98%
2-(Cyclopropylamino)ethanol
Formel: C₅H₁₁NO
Storage Temperature: Omgivande
MDL Number: MFCD09815646
CAS nummer: 35265-06-6