1-(3-Methoxyphenyl)ethanol 97%
Leverantör: Apollo Scientific
OR953341-10G OR953341-25G OR953341-5G
APOSOR953341-10GEA 3250 SEK
APOSOR953341-10G APOSOR953341-25G APOSOR953341-5G
1-(3-Methoxyphenyl)ethanol 97%
1-(3-Methoxyphenyl)ethanol
Formel: C₉H₁₂O₂
MW: 152,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00021866
CAS nummer: 23308-82-9