2-(4-Pyridyl)propan-2-amine 95%
Leverantör: Apollo Scientific
OR953315-100MG OR953315-1G OR953315-250MG
APOSOR953315-100MGEA 2150 SEK
APOSOR953315-100MG APOSOR953315-1G APOSOR953315-250MG
2-(4-Pyridyl)propan-2-amine 95%
2-(4-Pyridyl)propan-2-amine
Formel: C₈H₁₂N₂
MW: 136,194 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08729286
CAS nummer: 566155-76-8